Learn More
Apexbio Technology LLC Gatifloxacin, 1g Cas# 112811-59-3
SDP
Supplier:  Apexbio Technology LLC A13131000
Encompass_Preferred
Fluoroquinolone antibiotic. Inhibits bacterial type II topoisomerases (IC50 values are 0.109 and 13.8 g/ml for E.coli DNA gyrase and S.aureus topoisomerase IV respectively). Displays potent activity against gram-positive and Other sizes are available. Please inqury us for quote.
Catalog No. 50-199-8508