Learn More
Medchemexpress LLC Gatifloxacin hydroch 5g | 121577-32-0 | 5G
SDP
Supplier:  Medchemexpress LLC HY10581A5G
Encompass_Preferred
Gatifloxacin hydrochloride (AM-1155 BMS-206584 PD135432) is a potent fluoroquinolone antibiotic with broad-spectrum antibacterial activity Gatifloxacin hydrochloride inhibits bacterial type II topoisomerases (IC50 13 8 g/ml for S aureus topoisomerase IV) and E coli DNA gyrase (IC50 0 109 g/ml)[1 Gatifloxacin hydrochloride can be used to treat bacterial conjunctivitis in vivo
Catalog No. 50-002-43725