Learn More
Med Vet International Non-Sterile Gauze Sponges, 3x3, 12-ply
Supplier:  Med Vet International NON25312H
Encompass_Preferred
Medline 100% Cotton Woven Gauze Sponges: These USP Type VII cotton sponges are ideal for wound dressings, wound packing and general wound care. Our strictly controlled manufacturing process assures clean, debris-free packing with folded edges to prevent unraveling. Non-Sterile 3" x 3", 12-ply.
Catalog No. 50-209-1983