Learn More
Cayman Chemical GavestInlsodIum salt 10mg
SDP
Supplier:  Cayman Chemical 3284510 MG
Encompass_Preferred
An NMDA receptor antagonist; selectively binds to the glycine site of the NMDA receptor over the glutamate binding site of NMDA, AMPA, or GluK receptors (Kis = 0.003, 10, 7.94, and 3.98 µM, respectively); selective for the NMDA receptor over a panel of 70 receptors (IC50s = >10 µM for all); reduces infarct volume in a rat model of MCAO-induced focal ischemia at 3 mg/kg
Catalog No. 50-294-6695