Learn More
Glass Expansion GazFit Connectors for 4mm OD side arm (PKT 4)
SDP
Supplier:  Glass Expansion GAZ04
Encompass_Preferred
GazFit Connectors for 4mm OD torch side arms (PKT 4). Compatible with Glass Expansion’s D-Torch (30-808-3910) for Shimadzu ICPE 9000/9800 ICP-OES, and for Glass Expansion’s Semi Demountable Torch (30-808-4335) for Shimadzu ICPS 7500/8100 and ICPE 9000/9800 ICP-OES. Makes connection to torch side arms with soft walled tubing for argon gas supply.Also compatible with Glass Expansion Laser Ablation Adaptors 21-809-4140, 31-808-3045.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-270-2986