Learn More
Gold Biotechnology Inc GB10B ProTM Electro E coli
SDP
Supplier:  Gold Biotechnology Inc CC20110X50
Encompass_Preferred
GoldBio s GB10B-Pro Electrocompetent E coli cells are equivalent to DH10B cells and are especially designed for the most demanding cloning applications GB10B-Pro cells will provide the greatest number of transformants for when your research requires it including assembling large and multi-DNA fragments cloning large ( 10 kb up to 350 kb) or difficult construct transformations working with synthetic bio-applications and even BAC cloning
Catalog No. 50-005-62337