Learn More
Gold Biotechnology Inc GB10BTM Electro E coli Cells
SDP
Supplier:  Gold Biotechnology Inc CC20010X100
Encompass_Preferred
GoldBio s GB10B Electrocompetent E coli cells are equivalent to DH10B cells GB10B cells are high efficiency 2 x 10 8 cfu/ g plasmid DNA suitable for a wide variety of applications requiring high transformation efficiencies such as cloning and subcloning as well as working with either cDNA or gDNA library construction GB10B Electrocompetent E coli cells have multiple features including the 80lacZ M15 marker which provides -complementation of the -galactosidase gene with blue/white screening protocol These cells also have the mcrA genotypic marker and the mcrBC mrr deletion which allows for cloning of DNA that contains methylcytosine and methyladenine
Catalog No. 50-005-62335