Learn More
Medchemexpress LLC GB111-NH2 hydrochloride | 99.26% | 610.14 | 1 ML

Supplier: Medchemexpress LLC HY115454A10MM/1ML
GB111-NH2 hydrochloride is a cysteine cathepsin inhibitor, which can be used for cancer study. It is for research use only and not sold to patients.
- Purity: 99.26%
- Molecular weight: 610.14
- Formula: C33H40ClN3O6
- Appearance: Solid
- Color: White to off-white
- Storage: -20°C, sealed storage, away from moisture and light. In solvent: -80°C, 6 months; -20°C, 1 month (sealed storage, away from moisture and light).
- Solubility (in vitro): DMSO: 125 mg/mL (204.87 mM; needs ultrasonic; hygroscopic DMSO has a significant impact on solubility, use newly opened DMSO).
- Related classifications: Metabolic enzyme/protease, cathepsin.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.