Learn More
Medchemexpress LLC GB111-NH2 hydrochloride | 99.3% | 610.14 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY115454A
Encompass_Preferred
GB111-NH2 hydrochloride is identified as a cysteine cathepsin inhibitor and is suitable for cancer studies. It is intended for research use only and is not sold to patients.
  • Cysteine cathepsin inhibitor
  • Can be used for cancer study
  • Available in solid and solution forms
Catalog No. 50-004-39299