Learn More
Gold Biotechnology Inc GB5 alphaTM Comp E coli Cells
SDP
Supplier:  Gold Biotechnology Inc CC10110X50
Encompass_Preferred
GoldBio s GB5-alpha Chemically Competent E coli cells are equivalent to DH5-alpha cells and are high efficiency cells suitable for transformation in a wide variety of routine applications such as plasmid isolation cloning and subcloning
Catalog No. 50-005-62235