Learn More
Aviva Systems Biology Corp GBA3 Antibody - N-terminal region (ARP57505_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57505_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GBA3. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human GBA3. Alternate names: CBGL1, GLUC, KLrP, MGC104276, MGC126878ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Pig, Rabbit, Rat
Catalog No. 50-286-325