Learn More
Cayman Chemical GBR 12909hydrochlorIde 5mg
SDP
Supplier:  Cayman Chemical 156225 MG
Encompass_Preferred
A potent inhibitor that blocks dopamine uptake (IC50 = 1-51 nM); effectively inhibits dopamine uptake in vivo, leading to consequent stimulation of dopamine receptors.
Catalog No. 50-297-1648