Learn More
Cayman Chemical GBR 13069hydrochlorIde 10mg
SDP
Supplier:  Cayman Chemical 3890310 MG
Encompass_Preferred
A dopamine uptake inhibitor; selectively inhibits dopamine uptake over norepinephrine uptake in rat occipital cortex slices (IC50s = 40 and 800 nM, respectively); induces ipsilateral rotation and increases locomotion in rats at 2.5, 5, and 10 mg/kg; prevents 2’-CH3-MPTP-induced decreases in neostriatal levels of dopamine, DOPAC, and HVA in mice at 5 mg/kg
Catalog No. 50-291-8909