Learn More
Medchemexpress LLC GBT1118 | 1628799-51-8 | C19H20N2O4 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY15318250MG
Encompass_Preferred
GBT1118 is a potent and orally active allosteric modifier of hemoglobin oxygen affinity, enhancing tolerance to severe hypoxia. It works by covalently and reversibly binding to the N-terminal valine of the hemoglobin α chain, allosterically increasing intracellular hemoglobin's affinity for oxygen. This action helps protect red blood cells from damage during severe hypoxia.
  • Potent and orally active allosteric modifier of hemoglobin oxygen affinity
  • Increases tolerance to severe hypoxia
  • Binds covalently and reversibly to the hemoglobin α chain
  • Increases intracellular hemoglobin affinity for oxygen
  • Protects red blood cells from damage during severe hypoxia
  • Improved oxygen delivery during hypoxia in animal studies
  • Increased blood oxygen loading in the lungs in animal studies
  • Decreased hypoxia in vital tissues and lowered lactate levels in animal studies
Catalog No. 50-003-74050