Learn More
Diagnostic Biosystems GCDFP-15 GCDFP-15
Supplier:  Diagnostic Biosystems PDM261
Encompass_Preferred
It recognizes a protein of 15kDa, identified as Gross cystic disease fluid protein 15 (GCDFP-15). It is a major protein component of benign breast gross cysts. It is a known marker of breast cancer, asit isfound in approximately 50% of all breast cancer specimens. GCDFP-15, also known as PIP, for prolactin inducible protein, is a prolactin and androgen controlled protein. This antibody is useful in the identification of metastatic breast carcinoma, or fluid analysis.
Catalog No. 50-240-7413