Learn More
Medchemexpress LLC GCN2-IN-7 | 2396465-33-9 | C22H23BrN8OS | 1 MG
SDP
Supplier:  Medchemexpress LLC HY1514601MG
Encompass_Preferred
GCN2-IN-7 is an orally active and selective general control nonderepressible 2 (GCN2) inhibitor with an IC50 of 5 nM. It demonstrates anti-tumor activity in vivo and is intended for research use only.
  • Orally active and selective GCN2 inhibitor.
  • Exhibits an IC50 of 5 nM against GCN2.
  • Demonstrates anti-tumor activity in vivo.
  • Alleviates MDSC-related T cell suppression and restores T cell proliferation in vitro.
  • Shows robust target engagement in Balb/c mice.
  • Inhibits tumor growth in the LL2 syngeneic mouse model.
  • Soluble in DMSO (50 mg/mL).
  • Can be prepared in 10% DMSO, 90% Corn Oil for in vivo studies.
Catalog No. 50-003-76948