Learn More
ABclonal Technology GCNT3 Rabbit pAb
SDP
Supplier:  ABclonal Technology A1320920UL
Encompass_Preferred
This gene encodes a member of the N-acetylglucosaminyltransferase family. The encoded protein is a beta-6-N-acetylglucosamine-transferase that catalyzes the formation of core 2 and core 4 O-glycans on mucin-type glycoproteins.
Catalog No. 50-001-15388