Learn More
Advanced ImmunoChemical, Inc. Disialoganglioside GD2, Bovine, Lyophilized From Water Solution, 0.5mg
Supplier:  Advanced ImmunoChemical, Inc. 9IG6B0.5MG
Encompass_Preferred
This item is manufactured by HyTest, Ltd and distributed by Advanced ImmunoChemical, Inc. Antigens / Neuroscience / Disialoganglioside GD2, bovine. Source:Bovine brain tissue.Applications:Prepared from bovine GD1b ganglioside by enzymatic disintegration of the terminal galactose.Known as a specific cancer ganglioside. The antibodies to the GD2 inhibit cell attachment to various extracellular matrix proteins (collagen, vitronectin, laminin, fibronectin).Analysis:Purity more than 98 % by HPTLC. MW 1665Presentation:Lyophilized from water solution.Storage Conditions:Before and after reconstitution at -20 °C (between -15 and -30 °C allowed)Material safety note:This product is sold for research use only. Standard Laboratory Practices should be followed when handling this material.
Catalog No. 50-153-9825
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.