Learn More
Selleck Chemical LLC Taselisib (GDC 0032) 5mg 1282512-48-4 RG7604
SDP
Supplier:  Selleck Chemical LLC S71035MG
Encompass_Preferred
Taselisib (GDC 0032, RG7604) is a potent, next-generation ? isoform-sparing PI3K inhibitor targeting PI3K?/?/? with Ki of 0.29 nM/0.12 nM/0.97nM, >10 fold selective over PI3K?. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC1033437