Learn More
Apexbio Technology LLC GDC-0068 (RG7440), 1001264-89-6, MFCD22124514, 5 mg
SDP
Supplier:  Apexbio Technology LLC A30065
Encompass_Preferred
MF: C24H32ClN5O2, MW: 458, Solubility: 22.9mg/mL in DMSO, Chemical Name: (2S)-2-(4-chlorophenyl)-1-(4-((5R,7R)-7-hydroxy-5-methyl-6,7-dihydro-5H-cyclopenta(d)pyrimidin-4-yl)piperazin-1-yl)-3-(propan-2-ylamino)propan-1-one. GDC-0068 (RG7440) is a highly selective small molecule inhibitor of Akt with IC50s of 5, 18 and 8 nM for Akt1, Akt2 and Akt3.
Catalog No. 50-101-4687