Learn More
Apexbio Technology LLC GDC-0152 873652-48-3 100mg
SDP
Supplier:  Apexbio Technology LLC A4224100
Encompass_Preferred
GDC-0152 (CAS 873652-48-3) is a small-molecule antagonist of inhibitor of apoptosis proteins (IAPs) including ML-IAP XIAP cIAP1 and cIAP2 It binds to the BIR domain of ML-IAP and the BIR3 domains of XIAP cIAP1 and cIAP2 with reported Ki values of 14 nM 28 nM 17 nM and 43 nM respectively GDC-0152 promotes degradation of cIAP1 and induces caspase-3/7 activation resulting in reduced viability of breast cancer cells while sparing normal epithelial cells In cellular models GDC-0152 disrupts interactions between XIAP and caspase-9 and between IAPs and Smac supporting its utility in apoptosis-related cancer research
Catalog No. 50-000-97237