Learn More
Medchemexpress LLC GDC-0152 | 873652-48-3 | 99.7% | 498.64 | 100 MG

Supplier: Medchemexpress LLC HY13638100MG
GDC-0152 is a potent inhibitor of IAPs (Inhibitor of Apoptosis Proteins) that binds to the BIR3 domains of XIAP, cIAP1, cIAP2, and the BIR domain of ML-IAP. It activates caspases 3 and 7 and induces rapid degradation of cIAP1, making it suitable for apoptosis research.
- Binds to BIR3 domains of XIAP, cIAP1, cIAP2
- Binds to BIR domain of ML-IAP
- Activates caspases 3 and 7 in a dose- and time-dependent manner
- Induces rapid degradation of cIAP1
- Has moderate predicted hepatic clearance and plasma-protein binding
- For research use only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.