Learn More
Medchemexpress LLC GDC-0310 | 1788063-52-4 | 98.8% | 543.48 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY13908150MG
Encompass_Preferred
GDC-0310 is a selective acyl-sulfonamide Nav1.7 inhibitor, demonstrating an IC50 of 0.6 nM for hNav1.7. It exhibits an EC50 of 1.1 μM in vivo and a Ki of 1.8 nM for Nav1.7, showcasing improved Nav selectivity and ADME properties.
  • Selective acyl-sulfonamide Nav1.7 inhibitor
  • IC50 of 0.6 nM for hNav1.7
  • EC50 of 1.1 μM in vivo
  • Ki of 1.8 nM for Nav1.7
  • Improved Nav selectivity and ADME properties
  • Exhibits t1/2 values of 5 h in rat, 46 h in dog, and 4.4 h in cynomolgus monkeys
Catalog No. 50-003-51625