Learn More
Medchemexpress LLC Gdc-0310 | 1788063-52-4 | 98.8% | 543.48 g·mol⁻¹ | C25H29Cl2FN2O4S | 5 MG

Supplier: Medchemexpress LLC HY1390815MG
GDC-0310 is a selective acyl-sulfonamide inhibitor of the human voltage-gated sodium channel Nav1.7, reported with sub-nanomolar potency (IC50 = 0.6 nM). Supplied as a high-purity research compound, it is used in in vitro and in vivo pharmacology and electrophysiology studies; recommended solvent is DMSO.
- Sub-nanomolar potency for hNav1.7 (IC50 0.6 nM).
- High purity (~98.8%) suitable for pharmacology experiments.
- Solid research reagent with reported preclinical ADME and pharmacokinetic data.
- Soluble in DMSO for stock solution preparation.
- Molecular formula C25H29Cl2FN2O4S; molecular weight 543.48 g·mol⁻¹.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.