Learn More
Medchemexpress LLC GDC-0339 | 1428569-85-0 | 99.9% | 465.50 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY16976100MG
Encompass_Preferred
GDC-0339 is a potent, orally bioavailable, and well-tolerated pan-Pim kinase inhibitor, with Kis of 0.03 nM, 0.1 nM, and 0.02 nM for Pim1, Pim2, and Pim3, respectively. It is being explored as a potential treatment for multiple myeloma.
  • Potent, orally bioavailable, and well-tolerated pan-Pim kinase inhibitor.
  • Discovered as a potential treatment for multiple myeloma.
  • Cytostatic with an IC50 of 0.1 μM for MM.1S cells.
  • Induces a constellation of Pim downstream signaling events consistent with inhibition of Pim kinases.
  • Efficacious in RPMI8226 and MM.1S human multiple myeloma xenograft mouse models.
Catalog No. 50-003-93583