Learn More
Selleck Chemical LLC GDC-0623 5mg 1168091-68-6 G-868
SDP
Supplier:  Selleck Chemical LLC S75535MG
Encompass_Preferred
GDC-0623 (G-868) is a potent and ATP-uncompetitive MEK1 inhibitor with Ki of 0.13 nM. Phase 1. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8083