Learn More
Medchemexpress LLC HY-111484 5mg Medchemexpress, GDC-0927 CAS:1642297-01-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1114845MG
Encompass_Preferred
Medchemexpress, HY-111484 5mg GDC-0927 CAS:1642297-01-5 GDC-0927 (SRN-927) is a novel, potent, non-steroidal, orally bioavailable, selective estrogen receptor antagonist. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8880