Learn More
LUNA NANOTECH INC Genomic Dna Purification Plus Kit, 96-Well Plate Format, 384 Preps
Supplier:  LUNA NANOTECH INC NKP261384
Encompass_Preferred
PuroSPIN-96™ Genomic DNA Purification Plus Kit, 96-Well Plate Format, 384 Preps
The PuroSPIN-96™ Genomic DNA Purification Plus Kit is designed to purify genomic DNA from different biological samples such as bacteria, yeast, mammalian cells, blood, and animal tissue using 96-well silica membrane purification plates. The genomic DNA is released from the cells or tissue using Lysis Buffer  LB-GD1, Lysis Buffer LB-GD2, and Proteinase K. The DNA is then loaded onto silica-based purification plates and washed to remove protein, salt, and other contaminants. Finally, the purified DNA is eluted from the column into a clean 96-well elution plate in Elution Buffer EB, or nuclease-free water.
In comparison to our PuroSPIN-96™ Genomic DNA Purificatio Kit, the Plus kit adds an additional 96-well plate filtration step to remove the insoluble and non-digested fraction from the lysate before loading it onto the 96-well silica spin plate. This prevents cont
Catalog No. 50-256-6917