Learn More
LUNA NANOTECH INC Genomic Dna Purification Plus Kit, 50 Preps
Supplier:  LUNA NANOTECH INC NK26150
Encompass_Preferred
PuroSPIN™ Genomic DNA Purification Plus Kit, 50 Preps
PuroSPIN™ Genomic DNA Purification Plus Kit is designed to provide simple and easy-to-use method for extraction and purification of genomic DNA from Gram-positive and Gram-negative bacteria, yeast, mammalian cells, whole blood and animal tissues.
In comparison to our standard Genomic DNA Purificatio Kit, the Plus kit adds an additional filtration step that removes the insoluble and non-digested fraction from the lysate before loading it onto the silica spin column. This prevents contaminating materials from binding to and blocking parts of the silica membrane, leading to an increased genomic DNA yield and higher purity of the final product. This is particularly benficial when workign with large samples that contain a lot of starting material that needs to be digested.
The purification technology is based on nucleic acid binding to silica spin-columns, which allows efficient removal of protein
Catalog No. 50-256-6937