Learn More
Medchemexpress LLC GE11 | 875142-25-9 | 99.6% | 1540.68 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYP10128100MG
Encompass_Preferred
GE11 is an active peptide suitable for cancer studies and targeted drug delivery. It acts as a smart carrier for antitumor drugs by combining with colloidal drug delivery systems. As a potent EGFR ligand, this dodecapeptide specifically binds to EGFR with high affinity and selectivity, making it valuable for the diagnosis and targeted delivery of drugs in radiotherapy, gene therapy, and chemotherapy against EGFR-positive tumors.
  • Active peptide for cancer studies
  • Combines with colloidal drug delivery systems as smart carriers
  • Potent EGFR ligand with high affinity and selectivity
  • Used in diagnosis and targeted drug delivery
  • Applicable for radiotherapy, gene therapy, and chemotherapy against EGFR-positive tumors
Catalog No. 50-004-24052