Learn More
Enzo Life Sciences GEA 3162 (10mg). CAS: 144575-47-3
SDP
Supplier:  Enzo Life Sciences ALX430004M010
Encompass_Preferred
Water soluble nitric oxide donor and potent inhibitor of ADP-induced platelet aggregation in platelet rich plasma (PRP). Stimulates cGMP production in platelets, granulocytes and polymorphonuclear leukocytes. Vasodepressant. Alternative name: 1,2,3,4-Oxatriazolium, 5-amino-3-(3,4-dichlorophenyl)-, chloride. Purity: ≥97% (TLC). Solubility: Soluble in water, methanol or DMSO. Long Term Storage: -20°C.
Catalog No. 50-200-9128