Learn More
Medchemexpress LLC HY-108708 10mg Medchemexpress, GeA-69 CAS:2143475-98-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10870810MG
Encompass_Preferred
Medchemexpress, HY-108708 10mg GeA-69 CAS:2143475-98-1 GeA-69 is a selective, highly cell permeable allosteric inhibitor of poly-adenosine-diphosphate-ribose polymerase 14 (PARP14) targeting macrodomain 2, with a Kd of 2.1 µM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8617