Learn More
Selleck Chemical LLC Gedatolisib (PKI-587) 5mg 1197160-78-3 PF-05212384
SDP
Supplier:  Selleck Chemical LLC S26285MG
Encompass_Preferred
Gedatolisib (PF-05212384, PKI-587) is a highly potent dual inhibitor of PI3K?, PI3K? and mTOR with IC50 of 0.4 nM, 5.4 nM and 1.6 nM in cell-free assays, respectively. Phase 2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-6190