Learn More
Medchemexpress LLC HY-101588 100mg Medchemexpress, Gefapixant CAS:1015787-98-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY101588100MG
Encompass_Preferred
Medchemexpress, HY-101588 100mg Gefapixant CAS:1015787-98-0 Gefapixant is an orally active P2X3 receptor ( P2X3R ) antagonist with IC 50 s of ~30 nM versus recombinant hP2X3 homotrimers and 100-250 nM at hP2X2/3 heterotrimeric receptors. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2181