Learn More
Enzo Life Sciences Gefitinib-based PROTAC 3 (100mg). CAS: 2230821-27-7
SDP
Supplier:  Enzo Life Sciences ENZCHM2610100
Encompass_Preferred
Gefitinib-based PROTAC 3 is targeting on EGFR, consisting of Gefitinib as the warhead linked to VHL-recruiting ligand. Purity: ≥99%. Solubility: Soluble in DMSO (75 mg/mL). Insoluble in water. Long Term Storage: -20°C.
Catalog No. 50-201-3728