Learn More
Medchemexpress LLC Gefitinib-d3 | 1173976-40-3 | 98.4% | C22H21D3ClFN4O3 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY50895S21MG
Encompass_Preferred
Gefitinib-d3 is a deuterium-labeled form of Gefitinib, a potent and selective EGFR tyrosine kinase inhibitor. It effectively inhibits EGF-stimulated tumor cell growth and blocks EGFR autophosphorylation, also demonstrating antitumor activity through autophagy induction. This compound is suitable for use as a tracer or an internal standard for quantitative analysis methods like NMR, GC-MS, or LC-MS.
  • Potent and selective EGFR tyrosine kinase inhibitor
  • Inhibits EGF-stimulated tumor cell growth and blocks EGFR autophosphorylation
  • Induces autophagy and exhibits antitumor activity
  • Can be used as a tracer
  • Ideal as an internal standard for quantitative analysis (NMR, GC-MS, LC-MS)
Catalog No. 50-003-57802