Learn More
Medchemexpress LLC Gefitinib impurity 2 | 246512-44-7 | MFCD26395622 | 99.6% | 309.37 g/mol | C15H23N3O4 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY10066310MM 1ML
Encompass_Preferred
Gefitinib impurity 2 is a characterized impurity of the EGFR tyrosine kinase inhibitor gefitinib, supplied as a high-purity analytical standard for research and quality-control use. Offered in both solid form and as a ready-to-use DMSO solution, it supports impurity profiling, assay validation, and analytical method development.
  • High purity suitable for analytical reference.
  • Available as solid or solution formats.
  • Supplied in DMSO for convenient reconstitution.
  • Stable under recommended storage conditions.
  • Suitable for impurity profiling and assay controls.
  • Includes verified molecular formula and CAS identifier.
Catalog No. 50-002-14206