Learn More
CPC Scientific H-Gly-Glu-Gly-Phe-Leu-Gly-DPhe-Leu-OH (trifluoroacetate salt) 25MG
SDP
Supplier:  CPC Scientific CATI001B
Encompass
Inhibitor of cathepsin D, can be used as an affinity ligand for the isolation of cathepsin D.ONE-LETTER SEQUENCE: GEGFLG-DPhe-LMOLECULAR FORMULA: C41H58N8O11MOLECULAR WEIGHT:838.96STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [61393-34-8], RESEARCH AREA: OsteoporosisREFERENCES: F.Gubensek et al., FEBS Lett., 71, 42 (1976)
Catalog No. 50-210-9103