Learn More
Genscript Corporation ToxinSensor™ Gel Clot Endotoxin Assay Kit
Catalog No. 50274481
Encompass
GenScript ToxinSensor™​ Gel Clot Endotoxin Assay Kit is designed as a simple and sensitive in vitro end-product endotoxin test for human and animal parenteral drugs, biological products, and medical devices. The LAL test is a qualitative test for Gram-negative bacterial endotoxin. Lyophilized Amebocyte Lysate as supplied in the kit is to be reconstituted with LAL Reagent Water and then mixed in equal parts with the solution being tested. After incubation, and in the presence of endotoxin, gelation occurs; in the absence of endotoxin, gelation does not occur. Lyophilized Amebocyte Lysate reagent is made from amebocyte lysate from the horseshoe crab (Tachypleus tridentatus).The protocol described herein conforms to those described in the FDA guideline. Similar performance requirements for gel clot assays have been published and are updated regularly in the United States Pharmacopeia.
Catalog No. 50-274-481 Supplier Genscript Corporation Supplier No. L00351
 Restricted Item
Add to Cart
Add to Cart
GenScript ToxinSensor™​ Gel Clot Endotoxin Assay Kit is designed as a simple and sensitive in vitro end-product endotoxin test for human and animal parenteral drugs, biological products, and medical devices. The LAL test is a qualitative test for Gram-negative bacterial endotoxin. Lyophilized Amebocyte Lysate as supplied in the kit is to be reconstituted with LAL Reagent Water and then mixed in equal parts with the solution being tested. After incubation, and in the presence of endotoxin, gelation occurs; in the absence of endotoxin, gelation does not occur. Lyophilized Amebocyte Lysate reagent is made from amebocyte lysate from the horseshoe crab (Tachypleus tridentatus).The protocol described herein conforms to those described in the FDA guideline. Similar performance requirements for gel clot assays have been published and are updated regularly in the United States Pharmacopeia.