Learn More
Cole-Parmer Cole-Parmer Gel-Loading Tips, 1 to 200 µL, Bulk Pack; 1000/Pk
Supplier:  Cole-Parmer 2571310
Encompass_Preferred
These polypropylene tips are rounded to fit easily into small instrument openings. Use these microcapillary tips for DNA sequencing, loading SDS PAGE gels and other critical sampling applications.
Catalog No. NC9884348