Learn More
Alkali Scientific Gel Loading Tips, 200uL 83mm Length, Round bore opening, 960 tips/Case, Sterile
SDP
Supplier:  Alkali Scientific RGL102ST
Encompass_Preferred
Gel Loading Tips, 200 µL, 83 mm Length are designed for precise and controlled sample loading in electrophoresis applications. The extra long 83 mm profile provides easy access to deep or narrow gel wells, while the round bore opening ensures smooth flow and minimizes bubble formation for accurate dispensing. Precision molded for consistent performance, these tips are ideal for DNA, RNA, and protein gel loading workflows. Packaged in 2 racks of 96 tips per inner pack, with 5 inner packs per case, totaling 960 tips per case. This high throughput packaging supports efficient laboratory use. Key Features: 200 µL volume capacity for large gel loading applications, 83 mm extra long length for easy access to deep or narrow wells, round bore opening for smooth dispensing, reduces bubble formation and sample loss, non filtered design for electrophoresis workflows, precision molded for consistent results, compatible with standard micropipettes.
Catalog No. 50-006-63163