Learn More
ILEX LIFE SCIENCES LLC GELACELL ALGND PLLA IN 24W PLT
SDP
Supplier:  ILEX LIFE SCIENCES LLC GC0805ALWP24A
Encompass_Preferred
Gelacell - Aligned PLLA scaffolds in 24-well plate for 3D cell culture (1 plate). INTRO: Gelacell, manufactured by Gelatex Technologies, is a unique, 3D microfibrous scaffold designed for advanced in vitro 3D cell culture and tissue engineering applications. DESCRIPTION: This product is a 24-well plate containing circular scaffolds made from pharmaceutical grade poly-L-lactide (PLLA) fibers spun in aligned manner and fixed to PET discs. This scaffold format offers straightforward visual imaging of cells directly from the well plate. Ideal for cultivation of complex systems like organoids, cell microenvironments, and tissue regeneration. The scaffold’s optimized design ensures thermal, chemical, and mechanical stability, while providing substantial swelling capabilities and porosity. The aligned fibers in the scaffold provide a structured pathway for cell orientation and growth (particularly beneficial for specialized cell cultures requiring directional tissue development).
Catalog No. 50-273-3925