Learn More
ILEX LIFE SCIENCES LLC GELACELL ALIGNED PLLA 10X10 CM
SDP
Supplier:  ILEX LIFE SCIENCES LLC GC0805ALSCC
Encompass_Preferred
Gelacell - Aligned PLLA 10x10 cm scaffold sheet for 3D cell culture. INTRO: Gelacell, manufactured by Gelatex Technologies, is a unique, 3D microfibrous scaffold designed for advanced in vitro 3D cell culture and tissue engineering applications. DESCRIPTION: This product is a 10x10 cm sheet with aligned fibers made from pharmaceutical grade poly-L-lactide (PLLA) that offers a 3D structure allowing cells to grow in a more physiologically relevant manner, promoting cell-cell interactions, tissue-like organization, and cell polarization. It can be cut into desired shape/sizes depending on experimental needs. The scaffold is ideal for cultivation of complex systems like cell microenvironments, tissue regeneration, and organoids. The aligned fibers provide a structured pathway for cell orientation and growth (particularly beneficial for specialized cell cultures requiring directional tissue development).
Catalog No. 50-273-3926