Learn More
ILEX LIFE SCIENCES LLC GELACELL ALIGNED PLLA INS 6W
SDP
Supplier:  ILEX LIFE SCIENCES LLC GC0805ALIN06A
Encompass_Preferred
Gelacell - Aligned PLLA scaffold inserts for 6-well plate 3D cell culture (6 pack). INTRO: Gelacell, manufactured by Gelatex Technologies, is a unique, 3D microfibrous scaffold designed for advanced in vitro 3D cell culture and tissue engineering applications. DESCRIPTION: Specially designed 6-well plate scaffold “inserts” with aligned fibers made from pharmaceutical grade poly-L-lactide (PLLA) offer culture conditions for short-to-medium durations. Perfectly tailored for cultivation of complex systems like cell microenvironments, tissue regeneration, and organoids. The optimized design ensures thermal, chemical, and mechanical stability, while also providing substantial swelling capabilities and porosity, allowing efficient nutrient diffusion and thereby preventing cellular waste build up. The aligned fibers in the scaffold provide a structured pathway for cell orientation and growth (particularly beneficial for specialized cell cultures requiring directional tissue development).
Catalog No. 50-273-3928