Learn More
ILEX LIFE SCIENCES LLC GELACELL GELATIN INSERT 6WELL
SDP
Supplier:  ILEX LIFE SCIENCES LLC GC0601RNIN06A
Encompass_Preferred
Gelacell - Gelatin scaffold inserts for 6-well plate 3D cell culture (6 pack). INTRO: Gelacell, manufactured by Gelatex Technologies, is a unique, 3D microfibrous scaffold specifically engineered for advanced in vitro 3D cell culture and tissue engineering applications. Gelacell offers exceptional biocompatibility and non-toxicity across many cell types. DESCRIPTION: Specially designed 6-well plate scaffold “inserts” made from pharmaceutical grade gelatin offer cell culture conditions for short-to-medium durations. Perfectly tailored for the cultivation of complex systems like cell microenvironments, tissue regeneration, and organoids, Gelacell also makes a robust platform for drug screening and other cell culture investigations. The optimized design of the scaffold ensures thermal, chemical, and mechanical stability, while also providing substantial swelling capabilities and porosity, allowing efficient nutrient diffusion and thereby preventing cellular waste build up.
Catalog No. 50-273-3948