Learn More
ILEX LIFE SCIENCES LLC GELACELL PLGA CELLCROWN 12WELL
SDP
Supplier:  ILEX LIFE SCIENCES LLC GC0800RNCC12B
Encompass_Preferred
Gelacell - PLGA scaffolds fixed to cell crowns in 12-well plate for 3D cell culture (1 plate). INTRO: Gelacell, manufactured by Gelatex Technologies, is a unique, 3D microfibrous scaffold specifically engineered for advanced in vitro 3D cell culture and tissue engineering applications. DESCRIPTION: This variant of Gelacell is specially designed for medium-to-long term cell culture durations. Scaffolds made from pharmaceutical grade poly lactic-co-glycolic acid (PLGA) are fixed to cell crowns in a 12-well plate. The cell crowns act as Boyden chambers for facilitating studies related to air-liquid interface culture, co-culture, complex tissue models, etc. Gelacell also makes a robust platform for drug screening and other cell culture investigations. The optimized design ensures thermal, chemical, and mechanical stability, while also providing substantial swelling capabilities and porosity, allowing efficient nutrient diffusion and thereby preventing cellular waste build up.
Catalog No. 50-273-3943