Learn More
ILEX LIFE SCIENCES LLC GELACELL PLLA CELLCROWN 12WELL
SDP
Supplier:  ILEX LIFE SCIENCES LLC GC0805RNCC12B
Encompass_Preferred
Gelacell - PLLA scaffolds fixed to cell crowns in 12-well plate for 3D cell culture (1 plate). INTRO: Gelacell, manufactured by Gelatex Technologies, is a unique, 3D microfibrous scaffold designed for advanced in vitro 3D cell culture and tissue engineering applications. Designed as a non-woven, highly porous scaffold, Gelacell offers exceptional biocompatibility and non-toxicity across a variety of cell types. DESCRIPTION: This variant of Gelacell is specially designed for medium-to-long term culture durations. Scaffolds made from pharmaceutical grade poly-L-lactide (PLLA) are fixed to cell crowns in a 12-well plate. The cell crowns act as Boyden chambers for facilitating studies related to air-liquid interface culture, co-culture, complex tissue models, etc. The optimized design ensures thermal, chemical, and mechanical stability, while also providing substantial swelling capabilities and porosity, allowing efficient nutrient diffusion and thus preventing cellular waste build up.
Catalog No. 50-273-3923