Learn More
ILEX LIFE SCIENCES LLC GELACELL PLLA INSERTS 12-WELL
SDP
Supplier:  ILEX LIFE SCIENCES LLC GC0805RNIN12A
Encompass_Preferred
Gelacell - PLLA scaffold inserts for 12-well plate 3D cell culture (12 pack). INTRO: Gelacell, manufactured by Gelatex Technologies, is a unique, 3D microfibrous scaffold specifically engineered for advanced in vitro 3D cell culture and tissue engineering applications. Designed as a non-woven, highly porous scaffold, Gelacell offers exceptional biocompatibility and non-toxicity across a variety of cell types. DESCRIPTION: Specially designed 12-well plate scaffold “inserts” made from pharmaceutical grade poly-L-lactide (PLLA) offer cell culture conditions for short-to-medium durations. Perfectly tailored for the cultivation of complex systems like cell microenvironments, tissue regeneration, and organoids. The optimized design of the scaffold ensures thermal, chemical, and mechanical stability, while also providing substantial swelling capabilities and porosity, allowing efficient nutrient diffusion and thereby preventing cellular waste build up.
Catalog No. 50-273-3920