Learn More
Electron Microscopy Sciences Gelatin Powder Type B 75 Bloom, 3.5-5.0 (1% in H2O) pH 100 GR
SDP
Supplier:  Electron Microscopy Sciences 16566
Encompass_Preferred
F.W. 60000
Type A is from Porcine skin (acid cured tissue). Type B is from Bovine skin (base cured tissue). The strength of the gel is based on the bloom number. The higher the bloom number the stronger the gel. Used as a combination Acrylamide-Gelatin-Jung resin for cryosectioning of large and soft tissues (eggs, fishes, insects) Hartmann, R. (1984). A new embedding medium for cryosectioning eggs of high yolk and lipid content. Eur. J. Cell Biol., 34:206 Used to From Bovine skin (base cured tissue).
pH: 3.5-5.0 (1% in H2O)
Catalog No. 50-260-88