Learn More
Key Scientific Products Gelatin Test Strips, 50 per Pack
SDP
Supplier:  Key Scientific Products K250
Encompass_Preferred
GELATIN STRIPS, 50/PACK, Usage requires preliminary growth on media appropriate for the specime, Store tightly covered in a cool, dry place, The enzyme gelatinase, which liquifies gelatin, is one of these
Catalog No. NC9560006